Z-Phg-OH ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: Z-L-苯甘氨酸

C2567-1G C2567-5G
TCIAC2567-1GEA 0 CNY
TCIAC2567-1G TCIAC2567-5G
Z-Phg-OH ≥98.0% (by HPLC, titration analysis)
Z-Phg-OH
分子式: C₁₆H₁₅NO₄
分子量: 285.3 g/mol
MDL编号: MFCD00077033
CAS号: 53990-33-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D +114 to +118 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR