5-氯-2-(甲硫基)嘧啶-4-羧酸 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

C2549-1G C2549-5G
TCIAC2549-1GEA 0 CNY
TCIAC2549-1G TCIAC2549-5G
5-氯-2-(甲硫基)嘧啶-4-羧酸 ≥97.0% (by GC, titration analysis)
5-氯-2-(甲硫基)嘧啶-4-羧酸
分子式: C₆H₅ClN₂O₂S
分子量: 204.6300 g/mol
溶点: 170°C
MDL编号: MFCD00173907
CAS号: 61727-33-1

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR