Ciprofloxacin ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

C2510-25G C2510-5G
TCIAC2510-25GEA 0 CNY
TCIAC2510-25G TCIAC2510-5G
Ciprofloxacin ≥98.0% (by HPLC, titration analysis)
Ciprofloxacin
分子式: C₁₇H₁₈FN₃O₃
分子量: 331.35 g/mol
溶点: 265°C
MDL编号: MFCD00185755
CAS号: 85721-33-1
Merck Index: 14,02314

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR