Clofarabine ≥98.0% (通过HPLC)

供应商: TCI

C2500-100MG C2500-20MG
TCIAC2500-100MGEA 0 CNY
TCIAC2500-100MG TCIAC2500-20MG
Clofarabine ≥98.0% (通过HPLC)
Clofarabine
分子式: C₁₀H₁₁ClFN₅O₃
溶点: 227°C
存储条件: Refrigerator
MDL编号: MFCD00871077
CAS号: 123318-82-1
Merck Index: 14,02372

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR