Carnosic acid ≥97.0% (通过HPLC,滴定分析)

供应商: TCI

C2488-100MG C2488-20MG
TCIAC2488-100MGEA 0 CNY
TCIAC2488-100MG TCIAC2488-20MG
Carnosic acid ≥97.0% (通过HPLC,滴定分析)
Carnosic acid
分子式: C₂₀H₂₈O₄
溶点: 190°C
存储条件: Freezer
MDL编号: MFCD02259459
CAS号: 3650-09-7

现在订购

产品技术规格

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D +182 to +195 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR