Chlorpromazine hydrochloride ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-Chloro-10-[3-(dimethylamino)-1-propyl]phenothiazine Hydrochloride
C2481-25G
C2481-5G
TCIAC2481-25GEA
0
CNY
TCIAC2481-25G
TCIAC2481-5G
Chlorpromazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Chlorpromazine hydrochloride
分子式:
C₁₇H₁₉ClN₂S 分子量: 355.32 g/mol |
MDL编号:
MFCD00012654 CAS号: 69-09-0 EINECS: 200-701-3 UN号: 2811 ADR: 6.1 I Merck Index: 14,02185 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...