Carbidopa monohydrate ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: (S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic Acid

C2450-1G C2450-100MG
TCIAC2450-1GEA 0 CNY
TCIAC2450-1G TCIAC2450-100MG
Carbidopa monohydrate ≥98.0% (by HPLC, titration analysis)
Carbidopa monohydrate
分子式: C₁₀H₁₄N₂O₄
分子量: 226.23 g/mol
MDL编号: MFCD00889211
CAS号: 38821-49-7

现在订购

产品技术规格

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Specific rotation [a]20/D -21.0 ~ -23.5 deg (C=1, AlCl3 sol.)
Drying loss 5.9 to 8.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR