3-环己烯-1-甲醇 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1,2,3,6-四氢苯甲醇, 环己烯-4-基甲醇

C2435-25G C2435-250G
TCIAC2435-25GEA 0 CNY
TCIAC2435-25G TCIAC2435-250G
3-环己烯-1-甲醇 ≥98.0% (by GC)
3-环己烯-1-甲醇
分子式: C₇H₁₂O
分子量: 112.17 g/mol
沸点: 100°C
闪点: 73°C
MDL编号: MFCD00001574
CAS号: 1679-51-2
EINECS: 216-847-6

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR