4-(2-Chloroethylsulphonyl)butyric acid ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
C2428-25G
C2428-5G
TCIAC2428-25GEA
0
CNY
TCIAC2428-25G
TCIAC2428-5G
4-(2-Chloroethylsulphonyl)butyric acid ≥97.0% (by GC, titration analysis)
4-(2-Chloroethylsulphonyl)butyric acid
分子式:
C₆H₁₁ClO₄S 分子量: 214.66 g/mol |
CAS号:
121315-24-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...