Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
SingaCycle(TM)-A2
C2406-1G
C2406-200MG
TCIAC2406-1GEA
0
CNY
TCIAC2406-1G
TCIAC2406-200MG
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](N,N-dimethyl-3,5-dimethoxybenzylamine)palladium(II)]
This product is unavailable in the U.S.
分子式:
C₃₈H₅₂ClN₃O₂Pd 分子量: 724.72 g/mol |
CAS号:
1093348-08-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...