α-环戊基扁桃酸 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-环戊基-2-羟基苯乙酸
C2394-25G
C2394-5G
TCIAC2394-25GEA
0
CNY
TCIAC2394-25G
TCIAC2394-5G
α-环戊基扁桃酸 ≥98.0% (by HPLC, titration analysis)
α-环戊基扁桃酸
分子式:
C₁₃H₁₆O₃ 分子量: 220.27 g/mol |
MDL编号:
MFCD00019296 CAS号: 427-49-6 EINECS: 207-047-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 146.0 to 150.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...