Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver ≥97.0% (by total nitrogen basis)
供应商: TCI
Synonyms:
(1,3-Bis(2,6-diisopropylphenyl)-1H-imidazol-2(3H)-ylidene)silver(III) chloride
C2373-1G
C2373-200MG
TCIAC2373-1GEA
0
CNY
TCIAC2373-1G
TCIAC2373-200MG
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver ≥97.0% (by total nitrogen basis)
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]silver
分子式:
C₂₇H₃₆AgClN₂ 分子量: 531.92 g/mol |
MDL编号:
MFCD15072152 CAS号: 873297-19-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...