6-Chloropyrimidin-4(1H)-one ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 6-Chloro-4(1H)-pyrimidinone, 6-Chloro-4-pyrimidinol, 4-Chloro-6-hydroxypyrimidine

C2368-1G C2368-5G
TCIAC2368-1GEA 0 CNY
TCIAC2368-1G TCIAC2368-5G
6-Chloropyrimidin-4(1H)-one ≥98.0% (by GC, titration analysis)
6-Chloropyrimidin-4(1H)-one
分子式: C₄H₃ClN₂O
分子量: 130.5324 g/mol
MDL编号: MFCD00233957
CAS号: 4765-77-9

现在订购

产品技术规格

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 192.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR