5-Chloro-2-hydroxynicotinic acid ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 5-Chloro-2-hydroxy-3-pyridinecarboxylic Acid

C2339-1G C2339-5G
TCIAC2339-1GEA 0 CNY
TCIAC2339-1G TCIAC2339-5G
5-Chloro-2-hydroxynicotinic acid ≥98.0% (by titrimetric analysis)
5-Chloro-2-hydroxynicotinic acid
分子式: C₆H₄ClNO₃
分子量: 173.55 g/mol
溶点: 262°C
MDL编号: MFCD00204158
CAS号: 38076-80-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR