5-氯-1-甲基咪唑 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

C2317-25G C2317-5G
TCIAC2317-25GEA 0 CNY
TCIAC2317-25G TCIAC2317-5G
5-氯-1-甲基咪唑 ≥98.0% (by GC, titration analysis)
5-氯-1-甲基咪唑
分子式: C₄H₅ClN₂
分子量: 116.55 g/mol
沸点: 85°C
闪点: 96°C
MDL编号: MFCD00014505
CAS号: 872-49-1
EINECS: 212-827-6

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR