4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide ≥98.0% (by HPLC, total nitrogen)

供应商: TCI

Synonyms: Bezafibrate EP Impurity A, N-(4-Chlorobenzoyl)tyramine, 4-Chloro-N-(4-hydroxyphenethyl)benzamide

C2309-5G C2309-1G
TCIAC2309-5GEA 0 CNY
TCIAC2309-5G TCIAC2309-1G
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide ≥98.0% (by HPLC, total nitrogen)
4-Chloro-N-[2-(4-hydroxyphenyl)ethyl]benzamide
分子式: C₁₅H₁₄ClNO₂
分子量: 275.73 g/mol
MDL编号: MFCD00218513
CAS号: 41859-57-8
EINECS: 255-566-3

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR