氯[1,3-双(2,6-二异丙基苯基)咪唑-2-亚基]铜(I) ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: [1,3-双(2,6-二异丙基苯基)咪唑-2-亚基]氯化铜(I)

C2304-1G C2304-200MG
TCIAC2304-1GEA 0 CNY
TCIAC2304-1G TCIAC2304-200MG
氯[1,3-双(2,6-二异丙基苯基)咪唑-2-亚基]铜(I) ≥98.0% (by titrimetric analysis)
氯[1,3-双(2,6-二异丙基苯基)咪唑-2-亚基]铜(I)
分子式: C₂₇H₃₆ClCuN₂
MDL编号: MFCD09264276
CAS号: 578743-87-0

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR