2-Chloroethyl benzenesulphonate ≥98.0% (by GC)
供应商: TCI
Synonyms:
Benzenesulphonic Acid 2-Chloroethyl Ester
C2230-25G
C2230-5G
TCIAC2230-25GEA
0
CNY
TCIAC2230-25G
TCIAC2230-5G
2-Chloroethyl benzenesulphonate ≥98.0% (by GC)
2-Chloroethyl benzenesulphonate
分子式:
C₈H₉ClO₃S 分子量: 220.67 g/mol 沸点: 188°C |
CAS号:
16670-48-7 |
产品技术规格
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...