2,2'-O-Cyclocytidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

供应商: TCI

Synonyms: Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride

C2207-1G C2207-5G
TCIAC2207-1GEA 0 CNY
TCIAC2207-1G TCIAC2207-5G
2,2'-O-Cyclocytidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
2,2'-O-Cyclocytidine hydrochloride
分子式: C₉H₁₁N₃O₄
分子量: 261.66 g/mol
溶点: 264°C
MDL编号: MFCD00012636
CAS号: 10212-25-6
EINECS: 233-515-6
Merck Index: 14,00629

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -19.0 to -23.0 deg (C=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR