Carbon nanotube multi-walled, Length: 5 µm, Ø 60-100 nm
供应商: TCI
|
Warning
|
Synonyms:
CNT
C2158-5G
C2158-1G
TCIAC2158-5GEA
0
CNY
TCIAC2158-5G
TCIAC2158-1G
Carbon nanotube multi-walled, Length: 5 µm, Ø 60-100 nm
Carbon nanotube
分子式:
C 分子量: 12.011 g/mol |
MDL编号:
MFCD00133992 CAS号: 308068-56-6 |
产品技术规格
Appearance | Black powder to crystal |
Purity (TPO Method) | min. 95 % |
Diameter | 60 to 100 nm |
Length | min. 5 micro_m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...