trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥96.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
trans-4-(Cbz-amino)cyclohexanecarboxylic Acid, trans-4-(((Benzyloxy)carbonyl)amino)cyclohexanecarboxylic acid, trans-4-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
C2036-1G
C2036-25G
C2036-5G
TCIAC2036-1GEA
0
CNY
TCIAC2036-1G
TCIAC2036-25G
TCIAC2036-5G
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid ≥96.0% (by HPLC, titration analysis)
trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic acid
分子式:
C₁₅H₁₉NO₄ 分子量: 277.32 g/mol |
MDL编号:
MFCD01550737 CAS号: 34771-04-5 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...