1-Chloroethylcyclohexyl carbonate ≥98.0% (通过GC)

供应商: TCI
Warning

C1786-25G C1786-250G
TCIAC1786-25GEA 0 CNY
TCIAC1786-25G TCIAC1786-250G
1-Chloroethylcyclohexyl carbonate ≥98.0% (通过GC)
1-Chloroethylcyclohexyl carbonate
分子式: C₉H₁₅ClO₃
闪点: 84°C
MDL编号: MFCD04038149
CAS号: 99464-83-2
EINECS: 444-950-8
UN号: 1760
ADR: 8,II

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR