5-氯-2,4,6-三氟嘧啶 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
C1666-25G
C1666-5G
TCIAC1666-25GEA
0
CNY
TCIAC1666-25G
TCIAC1666-5G
5-氯-2,4,6-三氟嘧啶 ≥98.0% (by GC)
5-氯-2,4,6-三氟嘧啶
分子式:
C₄ClF₃N₂ 沸点: 115°C |
MDL编号:
MFCD00191933 CAS号: 697-83-6 EINECS: 211-807-4 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...