cis-6-Chloro-2-hexene ≥90.0% (通过GC)
供应商: TCI
|
Warning
|
C1653-25ML
C1653-5ML
TCIAC1653-25MLEA
0
CNY
TCIAC1653-25ML
TCIAC1653-5ML
cis-6-Chloro-2-hexene ≥90.0% (通过GC)
cis-6-Chloro-2-hexene
分子式:
C₆H₁₁Cl 沸点: 132°C |
MDL编号:
MFCD02093458 CAS号: 62614-70-4 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...