4-氯-1,2-二甲苯 (mixture of 3 and 4 isomers) ≥97.0% (通过GC)

供应商: TCI
Warning

C1626-25G C1626-500G
TCIAC1626-25GEA 0 CNY
TCIAC1626-25G TCIAC1626-500G
4-氯-1,2-二甲苯 (mixture of 3 and 4 isomers) ≥97.0% (通过GC)
4-氯-1,2-二甲苯
分子式: C₈H₉Cl
MDL编号: MFCD00000596
CAS号: 615-60-1

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR