卡托普利 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
Synonyms:
N-[(S)-3-巯基-2-甲基丙酰基]-L-脯氨酸
C1580-25G
C1580-5G
TCIAC1580-25GEA
0
CNY
TCIAC1580-25G
TCIAC1580-5G
卡托普利 ≥98.0% (by HPLC, titration analysis)
卡托普利
分子式:
C₉H₁₅NO₃S |
MDL编号:
MFCD00168073 CAS号: 62571-86-2 EINECS: 263-607-1 Merck Index: 14,01774 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Potassium iodate Method) | min. 98.0 % |
Melting point | 105.0 to 110.0 deg-C |
Specific rotation [a]20/D | -125 to -134 deg (C=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...