2-Chloro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)

供应商: TCI

C1560-1G C1560-100MG
TCIAC1560-1GEA 0 CNY
TCIAC1560-1G TCIAC1560-100MG
2-Chloro-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
2-Chloro-1,10-phenanthroline
分子式: C₁₂H₇ClN₂
分子量: 214.65 g/mol
MDL编号: MFCD00185678
CAS号: 7089-68-1

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR