N-(苄氧羰基)羟基胺 ≥97.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: N-Z-羟胺, N-羟基氨基甲酸苄酯

C1557-25G C1557-5G
TCIAC1557-25GEA 0 CNY
TCIAC1557-25G TCIAC1557-5G
N-(苄氧羰基)羟基胺 ≥97.0% (by HPLC, total nitrogen)
N-(苄氧羰基)羟基胺
分子式: C₈H₉NO₃
分子量: 167.16 g/mol
存储条件: Refrigerator
MDL编号: MFCD00010642
CAS号: 3426-71-9

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR