(S)-(+)-Camptothecin ≥97.0% (by HPLC)

供应商: TCI
Danger

Synonyms: (S)-4-Ethyl-4-hydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, (+)-Camptothecin, 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione

C1495-1G C1495-100MG
TCIAC1495-1GEA 0 CNY
TCIAC1495-1G TCIAC1495-100MG
(S)-(+)-Camptothecin ≥97.0% (by HPLC)
(S)-(+)-Camptothecin
分子式: C₂₀H₁₆N₂O₄
分子量: 348.36 g/mol
MDL编号: MFCD00081076
CAS号: 7689-03-4
UN号: 1544
ADR: 6.1 III
Merck Index: 14,01735

现在订购

产品技术规格

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +37.0 to +42.0 deg (C=0.4, CHCl3:MeOHl=8:2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR