2-氯-1,3-二甲基氯化咪唑啉 ≥98.0% (by HPLC)

供应商: TCI
Warning

Synonyms: DMC

C1408-25G C1408-5G
TCIAC1408-25GEA 0 CNY
TCIAC1408-25G TCIAC1408-5G
2-氯-1,3-二甲基氯化咪唑啉 ≥98.0% (by HPLC)
2-氯-1,3-二甲基氯化咪唑啉
分子式: C₅H₁₀Cl₂N₂
分子量: 169.05 g/mol
MDL编号: MFCD09039290
CAS号: 37091-73-9

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR