2-Ethylhexyl cyanoacetate ≥98.0% (通过GC)

供应商: TCI
Warning

C1384-25ML C1384-500ML
TCIAC1384-25MLEA 0 CNY
TCIAC1384-25ML TCIAC1384-500ML
2-Ethylhexyl cyanoacetate ≥98.0% (通过GC)
2-Ethylhexyl cyanoacetate
分子式: C₁₁H₁₉NO₂
MDL编号: MFCD00019843
CAS号: 13361-34-7
EINECS: 236-425-5

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR