(2S,3S)-2-Chloro-3-methylvaleric acid ≥95.0% (通过GC)

供应商: TCI
Warning

C1372-1G C1372-5G
TCIAC1372-1GEA 0 CNY
TCIAC1372-1G TCIAC1372-5G
(2S,3S)-2-Chloro-3-methylvaleric acid ≥95.0% (通过GC)
(2S,3S)-2-Chloro-3-methylvaleric acid
分子式: C₆H₁₁ClO₂
沸点: 125°C
MDL编号: MFCD00191403
CAS号: 32653-34-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Specific rotation [a]20/D -3.0 to -6.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR