5-Cyano-1H-imidazole-4-carboxamide hydrate ≥95.0% (by HPLC, titration analysis)

供应商: TCI
Danger

C1338-25G C1338-5G
TCIAC1338-25GEA 0 CNY
TCIAC1338-25G TCIAC1338-5G
5-Cyano-1H-imidazole-4-carboxamide hydrate ≥95.0% (by HPLC, titration analysis)
5-Cyano-1H-imidazole-4-carboxamide hydrate
分子式: C₅H₄N₄O
分子量: 136.11 g/mol
MDL编号: MFCD04038014
CAS号: 5372-23-6
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf927c4b24bf3406cb59bcd4075be928faa0d551&vid=b45e4904ab9b8487154b3f23f0cfc1b76be6b959&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0% (calcd.on anh.substance)
Solubility in hot Water almost transparency
Water max. 7.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR