4-Cyano-3-tetrahydrothiophenone ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Cyano-3-oxotetrahydrothiophene, 4-Oxotetrahydrothiophene-3-carbonitrile
C1164-25G
C1164-5G
TCIAC1164-25GEA
0
CNY
TCIAC1164-25G
TCIAC1164-5G
4-Cyano-3-tetrahydrothiophenone ≥98.0% (by GC)
4-Cyano-3-tetrahydrothiophenone
分子式:
C₅H₅NOS 分子量: 127.16 g/mol 沸点: 125°C |
MDL编号:
MFCD00059049 CAS号: 16563-14-7 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...