3-氯-4-羟基苯甲酸, hemihydrate ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

C1128-25G C1128-250G
TCIAC1128-25GEA 0 CNY
TCIAC1128-25G TCIAC1128-250G
3-氯-4-羟基苯甲酸, hemihydrate ≥98.0% (by titrimetric analysis)
3-氯-4-羟基苯甲酸
分子式: C₇H₅ClO₃½H₂O
分子量: 172.57 g/mol
沸点: 331
MDL编号: MFCD00002549
CAS号: 3964-58-7
EINECS: 223-574-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0% (calcd.on anh.substance)
Melting point 168.0 to 172.0 deg-C
Solubility in Methanol almost transparency
Water max. 5.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR