N-Cyclohexylethanolamine ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 2-(Cyclohexylamino)ethanol, N-(2-Hydroxyethyl)cyclohexylamine

C1082-25G C1082-500G
TCIAC1082-25GEA 0 CNY
TCIAC1082-25G TCIAC1082-500G
N-Cyclohexylethanolamine ≥98.0% (by titrimetric analysis)
N-Cyclohexylethanolamine
分子式: C₈H₁₇NO
分子量: 143.23 g/mol
MDL编号: MFCD00021326
CAS号: 2842-38-8
EINECS: 220-636-4

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR