2-氰乙基磷酸 ≥98.0% (通过滴定分析) phosphorylating agent

供应商: TCI
Danger

C0978-25G C0978-5G
TCIAC0978-25GEA 0 CNY
TCIAC0978-25G TCIAC0978-5G
2-氰乙基磷酸 ≥98.0% (通过滴定分析) phosphorylating agent
2-氰乙基磷酸
分子式: C₃H₆NO₄P
MDL编号: MFCD00012588
CAS号: 207121-42-4
UN号: 1564
ADR: 6.1,III

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water 5.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR