2-氰基-3,3-二苯基丙烯酸异辛酯 ≥98.0% (通过GC)

供应商: TCI

C0968-25G C0968-500G
TCIAC0968-25GEA 0 CNY
TCIAC0968-25G TCIAC0968-500G
2-氰基-3,3-二苯基丙烯酸异辛酯 ≥98.0% (通过GC)
2-氰基-3,3-二苯基丙烯酸异辛酯
分子式: C₂₄H₂₇NO₂
沸点: 218°C
溶点: –10°C
闪点: 203°C
MDL编号: MFCD00059260
CAS号: 6197-30-4
EINECS: 228-250-8
Merck Index: 14,06756

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR