1-(4-氯苯基)-3-甲基-2-吡唑啉-5-酮 ≥98.0% (通过滴定分析)

供应商: TCI

C0953-25G C0953-500G
TCIAC0953-25GEA 0 CNY
TCIAC0953-25G TCIAC0953-500G
1-(4-氯苯基)-3-甲基-2-吡唑啉-5-酮 ≥98.0% (通过滴定分析)
1-(4-氯苯基)-3-甲基-2-吡唑啉-5-酮
分子式: C₁₀H₉ClN₂O
MDL编号: MFCD00043814
CAS号: 13024-90-3
EINECS: 235-884-9

现在订购

产品技术规格

Purity (Neutralization titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR