1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate ≥96.0% (by total nitrogen and titration analysis) for peptide synthesis

供应商: TCI
Warning

Synonyms: CMC

C0793-25G C0793-5G
TCIAC0793-25GEA 0 CNY
TCIAC0793-25G TCIAC0793-5G
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate ≥96.0% (by total nitrogen and titration analysis) for peptide synthesis
1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimidemetho-p-toluenesulphonate
分子式: C₂₁H₃₃N₃O₄S
分子量: 423.57 g/mol
MDL编号: MFCD00011979
CAS号: 2491-17-0
EINECS: 219-650-3

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (with Total Nitrogen) min. 96.0 %
Purity (Ion exchange titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR