N-Carbobenzoxy-DL-norvaline ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
Z-DL-Nva-OH, N-Benzyloxycarbonyl-DL-norvaline, N-Cbz-DL-norvaline
C0752-25G
C0752-5G
TCIAC0752-25GEA
0
CNY
TCIAC0752-25G
TCIAC0752-5G
N-Carbobenzoxy-DL-norvaline ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-DL-norvaline
分子式:
C₁₃H₁₇NO₄ 分子量: 251.28 g/mol |
MDL编号:
MFCD00020434 CAS号: 21691-43-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 85.0 to 88.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...