Cholesteryl myristate ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Myristic acid Cholesterol ester, Cholesterol Myristate

C0675-10G C0675-25G
TCIAC0675-10GEA 0 CNY
TCIAC0675-10G TCIAC0675-25G
Cholesteryl myristate ≥98.0% (by titrimetric analysis)
Cholesteryl myristate
分子式: C₄₁H₇₂O₂
分子量: 597.025 g/mol
MDL编号: MFCD00014256
CAS号: 1989-52-2
EINECS: 217-867-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Ester Value) min. 98.0 %
Specific rotation [a]20/D -24.0 to -27.0 deg (C=2,chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR