3-环己烯-1-甲酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 1,2,3,6-四氢苯甲酸

C0652-25G C0652-500G
TCIAC0652-25GEA 0 CNY
TCIAC0652-25G TCIAC0652-500G
3-环己烯-1-甲酸 ≥98.0% (by GC, titration analysis)
3-环己烯-1-甲酸
分子式: C₇H₁₀O₂
分子量: 126.16 g/mol
沸点: 238°C
MDL编号: MFCD00013781
CAS号: 4771-80-6
EINECS: 225-314-7
UN号: 3265
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR