N-Carbobenzoxy-β-alanine ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Carbobenzyloxy-beta-alanine

C0640-1G C0640-10G
TCIAC0640-1GEA 0 CNY
TCIAC0640-1G TCIAC0640-10G
N-Carbobenzoxy-β-alanine ≥98.0% (by titrimetric analysis)
N-Carbobenzoxy-beta-alanine
分子式: C₁₁H₁₃NO₄
分子量: 223.23 g/mol
MDL编号: MFCD00037292
CAS号: 2304-94-1
EINECS: 218-967-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 107.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR