Z-DL-Val-OH ≥99.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
Z-DL-缬氨酸
C0634-1G
C0634-10G
TCIAC0634-1GEA
0
CNY
TCIAC0634-1G
TCIAC0634-10G
Z-DL-Val-OH ≥99.0% (by titrimetric analysis)
Z-DL-Val-OH
分子式:
C₁₃H₁₇NO₄ 分子量: 251.28 g/mol |
MDL编号:
MFCD00065127 CAS号: 3588-63-4 EINECS: 222-727-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 76.0 to 80.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...