Casein sodium salt (from milk)

供应商: TCI

C0594-500G C0594-25G
TCIAC0594-500GEA 0 CNY
TCIAC0594-500G TCIAC0594-25G
Casein sodium salt (from milk)
蛋白质

  • Total nitrogen: 14,5 to 15,8% (after drying)
  • Drying loss: Max. 12,0%
  • Ignition residue (Sulphate): Max. 6,0%

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR