Oleic acid cholesteryl ester ≥85.0% (by GC)

供应商: TCI

Synonyms: Oleic acid Cholesterol ester, Cholesteryl oleate, Cholesterol Oleate

C0559-25G C0559-500G
TCIAC0559-25GEA 0 CNY
TCIAC0559-25G TCIAC0559-500G
Oleic acid cholesteryl ester ≥85.0% (by GC)
Oleic acid cholesteryl ester
分子式: C₄₅H₇₈O₂
分子量: 651.12 g/mol
MDL编号: MFCD00003645
CAS号: 303-43-5
EINECS: 206-142-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 85.0 %
Specific rotation [a]20/D -18.0 to -22.0 deg (C=2, THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR