DL-Cystine mixture of DL/meso ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: (H-DL-Cys-OH)2

C0518-1G C0518-5G
TCIAC0518-1GEA 0 CNY
TCIAC0518-1G TCIAC0518-5G
DL-Cystine mixture of DL/meso ≥98.0% (by HPLC, titration analysis)
DL-Cystine
分子式: C₆H₁₂N₂O₄S₂
分子量: 240.3 g/mol
沸点: 429
溶点: 227
MDL编号: MFCD00084652
CAS号: 923-32-0
EINECS: 213-094-5
Merck Index: 14,02782

现在订购

产品技术规格

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 % (total of isomers)
Purity (Bromination Method) min. 98.0 %
Specific rotation [a]20/D -2.0 to +2.0 deg (C=1,1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR