Cyclohexylhydrazine hydrochloride ≥98.0% (通过滴定分析)

供应商: TCI
Warning

C0500-25G C0500-5G
TCIAC0500-25GEA 0 CNY
TCIAC0500-25G TCIAC0500-5G
Cyclohexylhydrazine hydrochloride ≥98.0% (通过滴定分析)
Cyclohexylhydrazine hydrochloride
分子式: C₆H₁₄N₂
MDL编号: MFCD00060160
CAS号: 24214-73-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR