Creatine phosphate disodium salt, hydrate ≥98.0% (通过HPLC)

供应商: TCI

C0397-1G C0397-25G
TCIAC0397-1GEA 0 CNY
TCIAC0397-1G TCIAC0397-25G
Creatine phosphate disodium salt, hydrate ≥98.0% (通过HPLC)
Creatine phosphate disodium salt
分子式: C₄H₈N₃O₅P·2Na
存储条件: Refrigerator
MDL编号: MFCD00014351
CAS号: 922-32-7
EINECS: 213-074-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Solubility in Water almost transparency
Water 22.0 to 30.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR